Order Entry
United States
ContactUsLinkComponent
µ-Dish 35 mm, High
µ-Dish 35 mm, High
Catalog # MSPP-81156
Supplier:  IBIDI USA MS
µ-Dish 35 mm, High
Catalog # MSPP-81156
Supplier:  IBIDI USA MS
Supplier Number:  81156

Specifications

  • Description:
    µ-Dish 35 mm, High
  • Size:
    2 ml
  • Cat. No.:
    MSPP-81156
  • Growth area:
    3.5 cm²
  • Supplier no.:
    81156
  • Details:
    Coating Area 4.1 cm² (w/400 µl)

Specifications

About this item

A 35 mm imaging dish with a polymer coverslip bottom for high-end microscopy and cell-based assays.

  • Equipped with a lid with locking feature that minimizes evaporation
  • Compaitable with a DIC Lid
  • Biocompatible plastic material—no glue, no leaking
  • Observation area of 21 mm

In this microscopy dish, the cells are imaged on a No. 1.5 ibidi Polymer Coverslip bottom with the highest optical quality and optimal cell adherence. This is a cell culture dish for everyday use with diverse application possibilities.