ContactUsLinkComponent
VWR® General Biology Dissecting Kit
VWR® General Biology Dissecting Kit
  76457-536
undefined
VWR® General Biology Dissecting Kit
  76457-536
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Dissection kit
  • Environmentally Preferable:
  • Cat. No.:
    76457-536
Wide variety of VWR® student dissection kits that suit educational needs.
Classroom and research dissecting applications.
 : ISO, GMP
 : Includes single fold vinyl case with lining (blue), #4 surgical handle with #21 replaceable blade; 5-1/2" sharp/blunt operating scissors; 5-1/2" straight thumb forceps (round, serrated tips; serrated grip); stainless steel mall-probe and seeker (hexagon 3¾" handle; 2½" seeker); straight teasing needle with 4" plastic handle; glass dropper with rubber bulb (large 5" size); 6" hard plastic ruler.