Specifications
- Description:Base buffer 1 mmol base/ml reagent
- Size:1 L
- Cat. No.:BDH85498.290
Specifications
About this item
The volumetric and coulometric reagents are buffered systems, stabilized around the ideal value between 5 to 7 ensuring a rapid and stoichiometric course of reaction. Buffering systems which neutralize and stabilize the pH during the reaction are suitable for this purpose.