VWR® Coulometric Reagents for Water Determination by the Karl Fischer Method
Supplier: VWR International
Certificates
About this item
Both anolyte and catholyte solutions are with improved formulation to increase the speed and accuracy of titration when determining water content at the microgram level.
- Reach endpoint quickly and consistently
- Long shelf life
- Stable endpoint
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.