ContactUsLinkComponent
SCIEX 4500 QTRAP Mass Spec System Package
SCIEX 4500 QTRAP Mass Spec System Package
Catalog # 76509-582
Supplier:  SCIEX
undefined
SCIEX 4500 QTRAP Mass Spec System Package
Catalog # 76509-582
Supplier:  SCIEX
Supplier Number:  5062218
Specifications
  • Description:
    4500 QTRAP mass spec system package
  • Cat. No.:
    76509-582
Specifications
About this item
Significantly bolster your confidence in your mass spec data. QTRAP technology is a triple quad system like no other, with all the performance of a triple quad but added functionality to overcome matrix interferences, increase screening throughput, and more.
  • The eQ™ Electronics and proven Qurved LINAC® Collision Cell provide unmatched support for fast chromatography applications with thousands of analytes
  • Turbo V Source: provides high-sensitivity analysis over a wide range of flow rates with quick-change APCI and TurboIonSpray® probes
  • QJet® Ion Guide: provides better collisional focusing to enhance ion transmission for ultimate sensitivity
  • AcQuRate Pulse Counting Detector: enables more accurate and precise ion detection over a wide dynamic range
QTRAP technology delivers equivalent or better data, and more of it, than you can capture on an ordinary triple quadrupole system. Overcome these 5 common challenges by taking the leap to QTRAP. Quantitation challenges due to complex matrices? QTRAP technology can decrease the worry for accurate quantitation, as it is free from matrix interferences. Increase data selectivity beyond triple quadrupole technology. The integrated Linear Ion Trap’s (LIT) unique MRM3 scan functionality enables quantitation from second generation fragment ions, decreasing matrix interferences without added sample prep.
Not sure about the accuracy of your compound ID? QTRAP technology can reduce the doubt for definitive compound identification. Increase specificity beyond triple quadrupole technology. QTRAP technology’s enhanced product ion (EPI) functionality gives you complete MS/MS spectra to cross reference with an integrated library for ultimate confirmation when reporting your results.
Looking to increase throughput by decreasing sample rechecks? QTRAP technology produces more high-quality results in every run. Increase throughput beyond triple quadrupole technology. By capturing MRM and enhanced product ion (EPI) MS/MS confirmation scans in one injection, you get high quality MRM quantitation with MS/MS confirmation at once for ultimate throughput and fewer rechecks.
Needing to find unknowns in your samples? QTRAP technology has the tools for your search. Screen for unknowns better than you can with triple quadrupole technology. With the enhanced mass spec (EMS), enhanced resolution (ER) and enhanced production (EPI) scanning capabilities, QTRAP can help you find and characterize components in your sample.
Being asked to do more than just quantitation? QTRAP technology gives you diversity. Enable your lab with functionality beyond triple quad technology. Sensitive and robust MRM quantitation combines with multiple other unique enhanced scan functions, giving you the quantitative performance of a triple quad system with added power to develop new methods or improve your existing workflows. Experience the power to do so much more on one mass spec system.
Certifications: NRTL certificate and EU Declaration of Conformity.
Delivery: 4500 QTRAP 4500 package includes enhanced high performance hybrid triple quadrupole/linear ion trap LC-MS/MS mass spectrometer, Turbo V Ion Source, One standard wet pump kit (roughing pump), Data Acquisition Workstation PC with Win10 64-bit OS, Standard Monitor and Standard parts and labour warranty for one year starting from the completion of instrument commissioning. Includes our StatusScope Remote Monitoring Service and one no-charge Preventative Maintenance (PM) during the one-year warranty period.
MS Hardware Additions
Product Information