SCIEX 4500 Triple Quad Mass Spec System Package
76509-720
: SCIEX
- Detector:AcQuRate Pulse Counting CEM
- Description:4500 Triple Quad mass spec system package
- Mass range:5 - 2000 Da
- Polarity switching:50 msec in MRM and Scheduled MRM modes
- Scan:Q1 MS, Q3 MS, Product Ion, Precursor Ion, Neutral Loss or Gain, MRM
- Cat. No.:76509-720
The Sciex Triple Quad 4500 System is a high sensitivity, bench top triple quadrupole mass spectrometer designed for LC-MS/MS analyses. This instrument provides excellent robustness and long term stability for the most demanding assays.
- The eQ™ Electronics and proven Qurved LINAC® Collision Cell provide unmatched support for fast chromatography applications with thousands of analytes
- Turbo V Source: provides high-sensitivity analysis over a wide range of flow rates with quick-change APCI and TurboIonSpray® probes
- QJet® Ion Guide: provides better collisional focusing to enhance ion transmission for ultimate sensitivity
- AcQuRate Pulse Counting Detector: enables more accurate and precise ion detection over a wide dynamic range
The 4500 system is a great fit for applications requiring round-the-clock quantitation, with the robustness to handle repeated injections of complex samples and reliability to minimize downtime. Such applications include food allergen testing, testing 93 forensic drugs in urine, amino acid quantitation, and gluten quantitation in foods.
: NRTL certificate and EU Declaration of Conformity.
: Can be field upgraded with QTRAP capabilities (76509-730).
: 4500 Triple Quad package includes enhanced high performance triple quadrupole LC-MS/MS mass spectrometer, Turbo V Source, TurboIonSpray (Heated Ion Spray) Probe, Atmospheric Pressure Chemical Ionization (APCI) Probe, One standard wet pump kit (roughing pump), Data Acquisition Workstation PC with Win10 64-bit OS, Standard Monitor and Standard parts and labour warranty for one year starting from the completion of instrument commissioning. Includes our StatusScope Remote Monitoring Service and one no-charge Preventative Maintenance (PM) during the one-year warranty period.