Orders LinkContactUsLinkComponent
Vi-CELL BLU Reagent Kits, Beckman Coulter
Vi-CELL BLU Reagent Kits, Beckman Coulter
Vi-CELL BLU Reagent Kits, Beckman Coulter
Supplier:  Beckman Coulter
Certificates
Vi-CELL BLU Reagent Kits, Beckman Coulter
Vi-CELL BLU Reagent Kits, Beckman Coulter
2 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
We have improved the Vi-CELL reagent pack to make it easier to use. The Vi-CELL reagents you know and trust are now packaged in a convenient ready-to-load kit. Simply open the instrument reagent panel and slide in the reagent pack. Each pack can be used for 310 activities before changing. Packs are easy to replace with no mess. Say goodbye to wrangling with tubes and splattering dye when you change out reagents.
Each pack also includes an RFID tag that tells you how many activities are remaining on the pack. No more guessing on remaining reagent amounts or running out in the middle of a critical series of tests.
The embedded RFID tag gives important information about each pack to your Vi-CELL BLU instrument: Lot number, expiration date, on-board life (90 days), activities remaining and part number.
Each reagent pack contains: Trypan blue bottle, waste bottle, buffer solution bottle, disinfectant bottle and cleaning agent bottle.
Ordering information: C06019 is for a single Vi-CELL BLU Reagent pack. C39291 is for a quad pack (Quantity 4) of Vi-CELL BLU reagent packs.
Documents
Product Family Options
Product Information
  • Availability
    Price
  • 76409-827
    Vi-CELL BLU reagent kit
    Vi-CELL BLU reagent kit
    Log in for availability
    Log in for availability
    Each
    Each - $572.00
    $572.00
    Each
       $572.00
    Supplier Number: C06019
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 76409-829
    Vi-CELL BLU quad pak (Quantity 4)
    Vi-CELL BLU quad pak (Quantity 4)
    Log in for availability
    Log in for availability
    Pack of 4
    Pack of 4 - $2,270.00
    $2,270.00
    Pack of 4
       $2,270.00
    Supplier Number: C39291
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.