- Description:Air compressor, Precision
- Cat. No.:103549-794
The Precision Air Compressor is designed and engineered specifically to provide a dedicated, fully integrated compressed air source for Precision Nitrogen and Precision Zero Air gas generators.
- Compressed air supply for Precision Nitrogen and Zero Air generators
- Single unit can supply both the Precision Nitrogen and Zero Air generators in a stack
- Duplex compressor mounting system to dampen noise and vibration
- Service and repair status indicators
- 12 month comprehensive on-site warranty
The Precision Air Compressor is designed and engineered specifically to provide a dedicated, fully integrated compressed air source for Precision Nitrogen and Precision Zero Air gas generators. Housed within the same compact, stackable form factor as the generators, the Precision Air Compressor module can be added to any Precision stack, typically as the base unit, to give a complete self-contained GC gas solution, even where no house supply is available.
Thanks to a unique duplex compressor mounting system, the Precision Air Compressor also benefits from low noise and vibration making it relatively quiet in operation within a typical laboratory.