ContactUsLinkComponent
Hydrogen Generator, Precision Hydrogen 300, Peak Scientific
Hydrogen Generator, Precision Hydrogen 300, Peak Scientific
Catalog # 103549-772
Supplier:  PEAK SCIENTIFIC MS
Irregular Voltage
undefined
Hydrogen Generator, Precision Hydrogen 300, Peak Scientific
Catalog # 103549-772
Supplier:  PEAK SCIENTIFIC MS
Supplier Number:  63-0300
Irregular Voltage
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Hydrogen generator, Precision Hydrogen 300
  • Cat. No.:
    103549-772
Specifications
About this item
The Precision Hydrogen generators are designed to provide the gas needed for detectors requiring hydrogen fuel gas, such as FID.
  • 99.9995% purity
  • Suitable for GC detector gas
  • Creates hydrogen on demand, minimal storage of hydrogen in the system
  • Low maintenance and minimal running costs over product lifetime
  • Internal leak detection with automatic shutdown
  • Automatic loading pump as standard
  • Simple maintenance, limited to replacing de-ionizer cartridge and silica gel
The Precision Hydrogen generators are designed to provide the gas needed for detectors requiring hydrogen fuel gas, such as FID. One generator is capable of supplying multiple detectors, and are available in various flow rates to suit individual customer needs.
These generators utilize a Proton Exchange Membrane (PEM) to create the hydrogen gas from deionized water and a desiccant filtration stage is used to dry the gas. The Precision Hydrogen generators are also suitable for providing collision gas for ICP-MS.
Precision Hydrogen gas generators come with various safety features as standard, giving you complete peace of mind in the laboratory and are a far safer, dependable and convenient alternative to cylinders.