ContactUsLinkComponent
Hydrogen Generator, Precision Hydrogen 100, Peak Scientific
Hydrogen Generator, Precision Hydrogen 100, Peak Scientific
Catalog # 103549-768
Supplier:  PEAK SCIENTIFIC MS
Irregular Voltage
undefined
Hydrogen Generator, Precision Hydrogen 100, Peak Scientific
Catalog # 103549-768
Supplier:  PEAK SCIENTIFIC MS
Supplier Number:  63-0100
Irregular Voltage
Specifications
  • Description:
    Peak, Scientific, Precision, Hydrogen 100, gas generator
  • Cat. No.:
    103549-768
Specifications
About this item
The Precision Hydrogen Standard generators are designed to provide the gas needed for GC detectors requiring hydrogen fuel gas, such as FID whilst the Precision Hydrogen Trace generators are capable of supplying GC carrier gas as well as detector gases for multiple GCs.
  • 99.9995% purity
  • Suitable for GC detector gas
  • Creates hydrogen on demand, minimal storage of hydrogen in the system
  • Low maintenance and minimal running costs over product lifetime
  • Internal leak detection with automatic shutdown
  • Automatic loading pump as standard
  • Simple maintenance, limited to replacing de-ionizer cartridge and silica gel
"The Precision Hydrogen Standard generators are designed to provide the gas needed for GC detectors requiring hydrogen fuel gas, such as FID whilst the Precision Hydrogen Trace generators are capable of supplying GC carrier gas as well as detector gases for multiple GCs. One generator is capable of supplying multiple detectors, and there are various flow rates available to suit individual laboratory needs.
These generators utilize a Proton Exchange Membrane (PEM) to create the hydrogen gas from deionized water and a desiccant filtration stage is used to dry the gas. The Precision Hydrogen generators are also suitable for providing collision gas for ICP-MS.
Precision Hydrogen gas generators come with various safety features as standard, giving you complete peace of mind in the laboratory and are a far safer, dependable and convenient alternative to cylinders.