ContactUsLinkComponent
Select Low Sulfur Columns, Agilent Technologies
Select Low Sulfur Columns, Agilent Technologies
Catalog # AGCP8575
undefined
Select Low Sulfur Columns, Agilent Technologies
Catalog # AGCP8575
Supplier Number:  CP8575
Specifications
  • Column Format:
    Capillary
  • Ø int.:
    0.32 mm
  • Description:
    GC column, 7" cage
  • Length:
    60 m
  • Environmentally Preferable:
  • Column hardware:
    Fused silica
  • Cat. No.:
    AGCP8575
  • Phase:
    Select Low Sulfur
Specifications
About this item
Deliver significantly lower detection limits for sulfur components in propylene streams.
  • Highest degree of column inertness provides better peak shape for active sulfur compounds
  • Low detection limits for sulfur compounds
  • Unique selectivity prevents co-elution and matrix interferences in propylene streams
  • Highly permeable PLOT stationary phase provides high retention of volatile components
  • Unique QC testing procedure results in consistent column inertness
  • Mechanical stability results in no particle loss
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W Select Low Sulfur meets the specific needs of petrochemical chemists who need lower detection limits for sulfur components in propylene. This column provides significantly lower detection limits due to the highest inertness of the highly permeable PLOT stationary phase towards volatile reactive sulfur components.