HP-PLOT Q Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Certificates
About this item
Designed to replace packed gas-solid columns and to separate ethane, ethylene, and ethyne (acetylene).
- Bonded polystyrene-divinylbenzene-based column
- A PLOT column with polarity between that of Porapak-Q and Porapak-N
- Excellent for C1 to C3 isomers and alkanes to C12, CO₂, methane, air/CO, oxygenated compounds, sulfur compounds, and solvents
- A PLOT column to replace packed gas-solid columns
- Separates ethane, ethylene, and ethyne (acetylene)
- Improved resolution in less time than conventional packed columns, with minimal conditioning time required (1 hour)
- Preferred 'Q' column due to its robust nature
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W HP-PLOT Q is a bonded polystyrene-divinylbenzene (DVB) column with polarity between Porapak-Q and Porapak-N. It was specially developed for the separation of targeted nonpolar and polar compounds, including hydrocarbons (natural gas, refinery gas, ethylene, propylene, all C1–C3 isomers); CO₂, methane, air/CO, and water; polar solvents (methanol, acetone, methylene chloride, alcohols, ketones, aldehydes, esters); and sulfur compounds (H₂S, mercaptans, COS).
Frequently Bought Together