J&W DB-Select 624 UI Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Certificates
About this item
Designed for residual solvent analysis of pharmaceutical compounds per US Pharmacopeia Method <467>.
- Engineered to optimize pharmaceutical residual solvents analysis per USP Method <467>
- Ultra inertness and low bleed
- Resolution of USP regulated critical pairs, also separates benzene and 1,2-dichloroethane
- Identical selectivity to the popular VF-624 ms – upgrade with no changes in method
- UI testing ensures premium performance column to column
- Tested against the strictest industry QC specifications for column bleed, sensitivity and efficiency
- Performance Summary shipped with each column
Agilent J&W DB-Select 624 UI <467> is designed specifically for United States Pharmacopoeia Method <467>. Equivalent to USP phase G43, this stationary phase is engineered for the best sensitivity and resolution of the critical pairs specified in <467> for the analysis of residual solvents in active pharmaceutical ingredients.
Specifications
- Catalog No:
- AG122-0334UI
- AG123-0364UI
- AG125-0334UI
- AG123-0334UI
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- USP Code:
- G43
- G43
- G43
- G43