DB-5 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Certificates
About this item
High-quality, nonpolar, general-purpose columns with low bleed and a high temperature limit.
- (5%-Phenyl)-methylpolysiloxane
- Nonpolar, low bleed and with a high temperature limit
- Bonded, crosslinked, and solvent rinsible
- Wide range of column dimensions available
- Excellent general purpose column with a wide range of applications
- Equivalent to USP Phase G27
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W DB-5 is nonpolar and low bleed and is available in a range of column dimensions. DB-5 is also bonded, crosslinked, and solvent-rinsible, and has a high temperature limit. It is equivalent to USP phase G27. The DB-5 column is tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency.
Specifications
- Catalog No:
- AGG3900-65006
- AG122-5012
- AG122-5032
- AG122-5033
- AG123-5034
- AG123-5033E
- AG123-5032-INT
- AG122-5063E
- AG128-5022
- AG123-5031
- AG122-5052
- AG128-5012
- AG123-502D
- AG123-5026
- AG122-5033LTM
- AG127-501E
- AG123-5022
- AG122-5032E
- AG123-5013E
- AG123-5012LTM
- AG122-5032-INT
- AG122-5031
- AG123-5011
- AG122-5022
- AG125-5032E
- AG125-5025
- AG122-5013
- AG122-503E
- AG123-5013
- AG123-5032E
- AG122-5061
- AG123-500ELTM
- AG123-5032
- AG123-5033
- AG121-5022
- AG123-5012
- AG122-5631
- AG125-501K
- AG121-5012
- AG125-501J
- AG121-5012E
- AG125-5017
- AG122-5062
- AG121-5042
- AG123-5063
- AG123-5062
- AG123-5053
- AG123-503ELTM
- AG125-50HB
- AG125-5012
- AG125-5035
- AG127-5012
- AG125-503J
- AG125-5065
- AG125-5065E
- AG122-5063
- AG123-503B
- AG125-5062
- AG123-503E
- AG123-5056
- AG125-503K
- AG125-503D
- AG125-503B
- AG125-5037
- AG125-5035E
- AG125-5034
- AG122-501E
- AG125-5032
- AG123-500E
- AG123-5003LTM
- AG122-5011
- AG127-5013
- AG122-5002LTM
- AG124-5037
- AG124-5032
- AG127-5023
- AG122-5632
- AG127-5022
- AG127-501N
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- USP Code:
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41