Anti-SARS-CoV-2 Human Monoclonal Antibody [clone: RV57]
Supplier: ReVacc Scientific
Certificates
About this item
This antibody has been tested to bind to receptor binding domain (RBD) of SARS-CoV-2 S protein, and show neutralizing activity against Wuhan strain of SARS-CoV-2 pseudovirus (Catalog # P14N41), but not Beta variant (Catalog # G99O57).
Specifications
- Catalog No:
- 76515-392
- 76515-394
- Antigen Symbol:
- SARS-CoV-2
- SARS-CoV-2
- Antigen Name:
- Spike, receptor binding domain
- Spike, receptor binding domain
- Antigen Synonyms:
- S, RBD
- S, RBD
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- RV57
- RV57
- Reactivity:
- Coronavirus
- Coronavirus
- Host:
- Human
- Human
- Gene ID:
- MN908947
- MN908947
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Modification:
- Recombinant Human Monoclonal Antibody
- Recombinant Human Monoclonal Antibody
- Epitope:
- Spike, receptor binding domain
- Spike, receptor binding domain
- Molecular Weight:
- 150 kDa
- 150 kDa
- Purification:
- Purified by Protein A chromatography
- Purified by Protein A chromatography
- Concentration:
- 1 mg/ml by Refractometry
- 1 mg/ml by Refractometry
- Storage Buffer:
- PBS, pH 7.2
- PBS, pH 7.2
- Storage Temperature:
- 4…–20 °C
- 4…–20 °C
- Shipping Temperature:
- Ice
- Ice
- Tested Applications:
- Dot Blot, FLISA
- Dot Blot, FLISA
Specifications
Frequently Bought Together