Order Entry
United States
ContactUsLinkComponent
 
Virus Recombinant Ebola Zaire Makona Trimeric GP (from Insect Cells)
 
undefined
Virus Recombinant Ebola Zaire Makona Trimeric GP (from Insect Cells)

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Virus
  • Storage Conditions:
    –80 °C
  • Gene ID:
    KR781609
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Ebola trimeric GP Δmuc protein-Zaire Makona strain (His and Strep Tags)
  • Protein/Peptide Name:
    Ebola zaire makona trimeric GP
  • Purity:
    60 - 85%
  • Molecular Weight:
    ~ 60 kDa
  • Endotoxin Level:
    None detected
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Frozen liquid. In PBS pH 7.5
  • Nuclease-free:
    N
  • Shipping Temperature:
    Dry Ice
  • Tested Applications:
    ELISA, SDS-PAGE

 

Frequently Bought Together