Virus Recombinant Dengue Virus 1 E Protein (from Insect Cells)
A recombinant form of envelope (E) protein from Dengue virus serotype 1, (GenBank: AAB70694) was produced by insect S2 cells, followed by purification.
- Protein function: Bind to its receptor
This product contains N-terminal Myc tag and C-terminal 6 hexa-histidine tag.
This product has been run on SEC (superose 6) and SDS-PAGE with reduced condition. The purity is not high. The he binding to Flavivirus control mAb 4G2 (targets domain II of E protein), and several published Dengue antibodies was tested in ELISAs.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Virus
- Storage Conditions:–80 °C
- Gene ID:AAB70694
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Dengue virus serotype 1 envelope protein (Myc and His Tags)
- Protein/Peptide Name:Dengue virus 1 E protein
- Purity:40 - 60%
- Molecular Weight:~ 45 kDa
- Endotoxin Level:None detected
- Concentration:0.2 mg/ml
- Formulation:Frozen liquid. In PBS pH 7.5
- Nuclease-free:N
- Shipping Temperature:Dry Ice
- Tested Applications:ELISA, SDS-PAGE, SEC
Frequently Bought Together