Order Entry
United States
ContactUsLinkComponent
Virus Recombinant SARS-COV-2 Trim Spike D614G Mutant 100 µg (from Mammalian Cells)
Virus Recombinant SARS-COV-2 Trim Spike D614G Mutant 100 µg (from Mammalian Cells)
Supplier:  ReVacc Scientific
Certificates
undefined
Virus Recombinant SARS-COV-2 Trim Spike D614G Mutant 100 µg (from Mammalian Cells)
Supplier:  ReVacc Scientific

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Mammalian cells
  • Species:
    Virus
  • Storage Conditions:
    –80 °C
  • Gene ID:
    MN908947
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    SARS-COV-2 trimeric spike protein-D614G mutant (His and Strep Tags)
  • Protein/Peptide Name:
    SARS-COV-2 trim spike D614G mutant
  • Purity:
    90 - 100%
  • Molecular Weight:
    ~ 140 kDa
  • Endotoxin Level:
    None detected
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Frozen liquid. In PBS pH 7.5
  • Nuclease-free:
    N
  • Shipping Temperature:
    Dry Ice
  • Tested Applications:
    ELISA, SDS-PAGE, NativeBlue, SEC

Specifications

Frequently Bought Together