Virus Recombinant SARS-COV-2 Trim Spike Wuhan Strain 100 µg (from Mammalian Cells)
Supplier: ReVacc Scientific
Certificates
About this item
A recombinant trimeric form of the spike glycoprotein (S protein) of SARS-Cov-2 (Wuhan strain, GenBank: MN908947) was produced by human embryonic kidney HEK293F cells, followed by purification.
- Protein function: Bind to its receptor
This product contains ectodomain of S protein with S1/S2 cleavage site knocked-down, and also had a pair of mutations (N1067P and L1068P), a HRV3C cleavage site, T4 foldon trimerization domain, C-terminal 8 hexa-histidine tag and twin-strep tag. Twin-strep tag can be used for streptavidin labeling.
This product has been run on SDS-PAGE with reduced condition and Native blue gels as well as SEC, with results proving trimerization and glycosylation. The binding to two control mAbs and CR3022 was tested in ELISAs.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:Virus
- Storage Conditions:–80 °C
- Gene ID:MN908947
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:SARS-COV-2 trimeric spike protein-Wuhan strain (His and Strep Tags)
- Protein/Peptide Name:SARS-COV-2 trim spike wuhan strain
- Purity:90 - 100%
- Molecular Weight:~ 140 kDa
- Endotoxin Level:None detected
- Concentration:0.2 mg/ml
- Formulation:Frozen liquid. In PBS pH 7.5
- Nuclease-free:N
- Shipping Temperature:Dry Ice
- Tested Applications:ELISA, SDS-PAGE, NativeBlue, SEC
Specifications
Frequently Bought Together