ContactUsLinkComponent
Capillary Column Installation Kit for GC, Agilent Technologies
Capillary Column Installation Kit for GC, Agilent Technologies
Catalog # AG430-2000
Capillary Column Installation Kit for GC, Agilent Technologies
Catalog # AG430-2000
Supplier Number:  430-2000
Specifications
  • Description:
    GC Column Installation Kit
  • Cat. No.:
    AG430-2000
Specifications
About this item
Agilent's GC capillary column installation kit includes a diamond-tipped pencil, 20x magnifier, ferrule and septum removal tools, and refillable two-ounce bottle of Snoop.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise.
The capillary column installation kit provides everything needed to easily install a GC capillary column. The kit includes a diamond-tipped pencil, 20x magnifier, ferrule removal tool, septum removal tool, and refillable two-ounce bottle of Snoop.