

Orthophosphoric acid 85% ACS, meets analytical specification of NF, VWR Chemicals BDH®
Catalog # BDH3118-3.8LP
Supplier: BDH
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Poly bottle
- Size:3.8 L
- Cat. No.:BDH3118-3.8LP
- Supplier no.:3118
Specifications
Specification Test Results
- Assay (H3PO4) :85.0-88.0%
- Alkali phosphates:To Pass test
- Identification: NF:To Pass test
- Limit of Nitrate:To Pass test
- Phosphorous or hypophosphorous acid:To Pass test
- Sulfate:To Pass test
- Manufacture Date :Record Actual
- Expiration Date:Record Actual
Specification Test Results
About this item
:
:98 g/mol
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
:GHS05
:GHS07
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340