ContactUsLinkComponent
Flowflex™ Covid-19 SARS COV-2 Antigen Home Test
Flowflex™ Covid-19 SARS COV-2 Antigen Home Test
Catalog # 76515-532
CAS Number:  
Flowflex™ Covid-19 SARS COV-2 Antigen Home Test
Catalog # 76515-532
Supplier Number:  L031-118B5
CAS Number:  
Specifications
  • Description:
    Flowflex™ COVID-19 Antigen Home Test
  • Cat. No.:
    76515-532
Specifications
About this item
Acon’s Flowflex™ COVID-19 Antigen Home Test is an over the counter test for COVID-19.
  • Nasal swab test
  • Quick results in 15 minutes
  • Excellent performance when compared to an FDA authorized molecular SARS-CoV-2 test
This product has been authorized only for the detection of proteins from SARS-CoV-2, not for any other viruses or pathogens.The emergency use of this product is only authorized for the duration of the declaration that circumstances exist justifying the authorization of emergency use of IVDs for detection and/or diagnosis of COVID-19 under Section 564(b)(1) of the Federal Food, Drug and Cosmetic Act, 21 U.S.C. § 360bbb-3(b)(1), unless the declaration is terminated, or authorization is revoked sooner.
Certifications: This product has been granted an EUA authorized by the FDA.
Ordering information: All sales are final for purchase and cannot be returned once received.