Order Entry
United States
ContactUsLinkComponent
Boletopsis grisea Lectin (BGL), New England Biolabs
Boletopsis grisea Lectin (BGL), New England Biolabs
Catalog # 76509-460
CAS Number:  
undefined
Boletopsis grisea Lectin (BGL), New England Biolabs
Catalog # 76509-460
Supplier Number:  P0867S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
  • Species:
  • Size:
    1 ml
  • Environmentally Preferable:
  • Cat. No.:
    76509-460

Specifications

About this item

Boletopsis grisea Lectin (BGL) is a recombinant 15 kDa lectin from the Boletopsis grisea mushroom that has been expressed in E. coli.

  • Enzyme for innovation
  • Binds to N-glycans having at least one terminal outer arm GlcNAc
  • Concentration: 1 mg/ml

Enables enrichment of GlcNAc-capped N-glycans or mucin type O-glycopeptides from complex samples in glycomics and glycoproteomics analytical workflows.