Specifications
- Sample Volume:10 µl
- Description:EVE™ Plus automated cell counter, US-plug
- Cell Size:5 - 60 µm
- Concentration:1×10⁴ - 2×10⁷ cells/ml
- Focus:Manual or automatic
- Measurement Time:1 s/test (manual focus), <10 s/test (auto focus)
- Cat. No.:76492-620
Specifications
About this item
EVE™ PLUS is a compact image-based automated cell counter that uses disposable slides to most accurately determine eukaryotic cell titers in less than 1 second. In addition, when applying trypan blue staining, the system will discriminate live from dead cells precisely to provide reliable data on cell viability. Compared to counting cells manually using classical hemocytometers the EVE™ PLUS automated cell counter offers three main benefits:
- Reproducible, objective data, free from human bias/error
- Quick and lean workflow saves times and increases productivity
- Disposable counting slides improve lab safety
Based on its well-established precursor EVE™ the EVE™ PLUS uses advanced optics and image analysis algorithms for counting even challenging clumped cell populations at a precision that is proven to be comparable to more complex fluorescence-based counting methods.
The cell concentration measuring range has been increased to 1×10⁴ to 2×10⁷ cells/ml with cell sizes ranging from 5 to 60 µm being recognized. The system can, therefore, be used on all kinds of round-shaped mammalian cell lines, primary cells, red blood cells, PBMC, and some round types of yeast. For irregular shaped cells we recommend you use the fluorescence-based cell counter ADAM-MC2.
A well-arranged touch screen user interface together with smart data storage and transfer options guarantee for easy and hassle-free operation.
EVE™ cell counting slides offer reproducible and reliable measurements and analysis. The unique design eliminates the use of cover slips and allows for exact volume control (10 µl cell suspension per test). In contrast to classical hemocytometers the disposable slides reduce the user’s risk of exposure to biological samples and eliminate the need for washing or autoclaving components. Each slide contains two enclosed chambers allowing for two tests per slide.
To prevent biased, unreliable results when using poor quality replicas, EVE™ slide packs do include an individual access code, which you must enter in the EVE™ PLUS software for authentication. Each access code is valid for up to 200 tests and can be used on an unlimited number of EVE™ PLUS instruments.