ContactUsLinkComponent
Semi-Automated Titrator, METTLER TOLEDO®
Semi-Automated Titrator, METTLER TOLEDO®
Catalog # 76307-656
Supplier:  Mettler Toledo
undefined
Semi-Automated Titrator, METTLER TOLEDO®
Catalog # 76307-656
Supplier:  Mettler Toledo
Supplier Number:  30470855
Specifications
  • Description:
    EasyPlus titrator Easy EPM
  • Burette Resolution:
    EasyPlus burette 20 ml (included)
  • Cat. No.:
    76307-656
  • Application:
    Potentiometric
Specifications
About this item
The EasyPlus Easy EPM performs precise semi-automated titrations with the automated burette, allowing the operator to focus on the color change of samples.
  • Precise semi-automated titrations
  • Error free results
  • Simple operation
  • iTitrate user interface
Perform titrations using the automated burette, focusing on the color change of samples. An immersed dispensing tip avoids drop by drop variations. A variety of calculations are already pre-programmed in the instrument. Simply select your measurement unit, and avoid error-prone manual calculations. Perform your analysis using either the touch screen or the new responsive USB wheel controller. Enjoy its precise control and comfortable operation.