Order Entry
United States
Orders LinkContactUsLinkComponent
Copper etchant CE-100 semiconductor grade

Copper etchant CE-100 semiconductor grade

Corrosive
Irritant
Health
Supplier:   Transene
Copper etchant CE-100 semiconductor grade
Supplier:  Transene
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Copper Etchant CE-100 is a fast etch for removal of thicker copper films.

Synonyms:  Copper etching solution
  • MDL Number:MFCD00161912
  • Density:1.3 g/cc
  • Storage Temperature:Ambient
  • UN:2582
  • ADR:8,III
Specifications for Product Family

Specification Test Results

  • Appearance:
    Brown
  • Etch Capacity (rate declines at ~70%):
    19 oz/gallon
  • Etch Rate at 40°C:
    1 (etches 1oz Cu in 90 sec) mL/minute
  • Filtration:
    1 um
  • Incompatible Materials:
    Al, Ni, Ti
  • pH:
    Acidic
  • Recommended Operating Temperatures:
    20-80°C (30-40°C most common) °C
  • Shelf Life:
    1 Year
  • Storage Conditions:
    Ambient
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • HDPE bottle
    Quart
    Log in for availability
    Log in for availability
    Each (1qt US)
    Each (1qt US) - $65.41
    $65.41
    Each (1qt US)
       $65.41

    Supplier Number: 006CE100-QT
  • HDPE bottle
    Gallon
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1US Gal)
    Each (1US Gal) - $147.67
    $147.67
    Log in for availability
    Case of 4 (1 US Gal)
    Case of 4 (1 US Gal) - $590.68
    $590.68
    Each (1US Gal)
       $147.67

    Case of 4 (1 US Gal)
       $590.68

    Supplier Number: 006CE100-GAL

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.