Order Entry
United States
ContactUsLinkComponent
Chromium etchant 1020 semiconductor grade
Corrosive
Irritant
Health
Chromium etchant 1020 semiconductor grade
Supplier:  Transene
Certificates
Corrosive
Irritant
Health
Chromium etchant 1020 semiconductor grade
Supplier:  Transene

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Appearance:
    Clear orange
  • Etch Capacity (rate declines at ~70%):
    65 g/gallon
  • Etch Rate at 40°C:
    40/sec Å/sec
  • Filtration:
    0,2 um
  • Incompatible Materials:
    Al, Ni, Cu, NiCr
  • pH:
    Acidic
  • Recommended Operating Temperatures:
    20-80°C (30-40°C most common) °C
  • Shelf Life:
    1 year
  • Storage Conditions:
    Ambient

Specification Test Results

Frequently Bought Together