Orders LinkContactUsLinkComponent
iFluor® 350, Tyramide
200 Slides
Catalog # 76485-376
Supplier:   AAT Bioquest
 :  45095
iFluor® 350, Tyramide
Catalog # 76485-376
Supplier:  AAT Bioquest
CAS Number:  
iFluor® 350, Tyramide
iFluor® 350, Tyramide
200 Slides
CAS Number:  
Price
$369.71
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
For many immunohistochemical (IHC) applications, the traditional enzymatic amplification procedures are sufficient for achieving adequate antigen detection.
  • increased fluorescence signal
  • suitable for low-abundance targets
  • compatible with all major fluorescence platforms
 : For research use only
Specifications
Specifications
  • Size:
    200 Slides