WB1600 Series Wrist Band and Coil Cord Set, Transforming Technologies
Catalog # 76490-244
Supplier: Transforming Technologies
Specifications
- Color:Blue
- Description:Fabric wrist band with 12' coil cord, 4 mm snap
- Cat. No.:76490-244
Specifications
About this item
The WB1600 Series Wrist Band and Coil Cord set is the most versatile and cost-effective band set offered by Transforming Technologies.
- Wrist strap is a durable and long-lasting method of grounding personnel
- Made with a soft woven fabric that is comfortable to wear for long periods
- Fabric band uses hypoallergenic silver-plated nylon thread for conductivity
- Band will not irritate skin
The soft, blue woven fabric and 360° curved buckle of the wrist band allows the skin to breathe and makes it very comfortable to wear. The band uses hypoallergenic silver-plated nylon thread for conductivity and has a maximum resistance of 160 ohms.