Order Entry
United States
ContactUsLinkComponent
6-Carboxy-2',4,7',7-tetrachlorofluorescein succinimidyl ester (6-TET SE) ≥90%
6-Carboxy-2',4,7',7-tetrachlorofluorescein succinimidyl ester (6-TET SE) ≥90%
Catalog # 76480-746
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
6-Carboxy-2',4,7',7-tetrachlorofluorescein succinimidyl ester (6-TET SE) ≥90%
Catalog # 76480-746
Supplier:  AAT BIOQUEST INC
Supplier Number:  211
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    5 mg
  • Cat. No.:
    76480-746

Specifications

About this item

 :
 :611.17 g/mol
Formula:C₂₅H₁₁Cl₄NO₉