Order Entry
United States
ContactUsLinkComponent
Propidium iodide 10 mM in aqueous solution ≥95%
Propidium iodide 10 mM in aqueous solution ≥95%
Catalog # 76482-862
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Propidium iodide 10 mM in aqueous solution ≥95%
Catalog # 76482-862
Supplier:  AAT BIOQUEST INC
Supplier Number:  17517
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 ml
  • Cat. No.:
    76482-862

Specifications

About this item

 :
 :668.4 g/mol
CAS Number:25535-16-4
Formula:C₂₇H₃₄I₂N₄
MDL Number:MFCD00011921