- Description:Texas Red® hydrazide
- Size:5 mg
- Purity:95%
- Cat. No.:76480-920
Texas Red® hydrazide reacts with carbonyl compounds such as aldehyde, ketones and acids to give bright red fluorescent conjugates.
- Identical spectra to Texas Red
- High Stability