BriskHeat® 55-Gallon Drum Cover, Antylia Scientific
Catalog # 76181-588
Supplier: Antyila Scientific
Specifications
- Description:BriskHeat® FGDC55 55 gal. drum insulation top cover, plastic
- Cat. No.:76181-588
- Fits:55 gal. drum
Specifications
About this item
Prevent heat loss from your drum heater.
- Suitable for use with the Briskheat® 55-gallon insulated drum heaters
This BriskHeat 55-gallon drum insulation top cover is used to minimize heat loss and speed up heat-up time. Made to fit standard 55-gallon drums with cut-outs to access plug and bung without removing cover.