EZ-Zyme® Instrument Cleaner Packets, Mortech®
EZ-Zyme® Instrument cleaner packets
Catalog # 76498-518
Supplier: Mopec
Supplier #: BE100
Jump to:
Product Details & Documents
This product is one of the best enzyme cleaners on the market. Although not a disinfectant, it does dissolve all tissue debris, leaving every crack and crevice of dissection instruments looking brand new.
- Contains 32 packets
Simply open the packet of concentrate, dilute into a gallon of warm tap water and place any hard-to-clean instrument into the solution. There is absolutely no hand scrubbing required.
Specifications
- Description:EZ-Zyme® Instrument cleaner packets