Orders LinkContactUsLinkComponent
PathOdorizer, Odor Eliminating Solution, Mortech®
PathOdorizer, Odor Eliminating Solution, Mortech®
Supplier:   Mopec
PathOdorizer, Odor Eliminating Solution, Mortech®
Supplier:  Mopec
Certificates
PathOdorizer, Odor Eliminating Solution, Mortech®
PathOdorizer, Odor Eliminating Solution, Mortech®
2 Options Available Below
Product Details & Documents
Developed specially for use by medical examiners, PathOdorizer eliminates offensive odors emitted from decomposed bodies within seconds. From the discovery scene to the morgue, PathOdorizer can be used in every phase of work.
  • No CFCs, formaldehyde, or hazardous biocides
  • No toxic fumes
  • Safe for use on metals, plastics, or fabrics
  • PathOdorizer Plus offers additional cleaning and degreasing properties
PathOdorizer not only removes odors from clothes, vehicles, body bags, refrigerators, and other items, it keeps the odor from permeating the lab environment. The formula is completely water soluble, odorless, colorless, biodegradable, non-flammable, and non-corrosive.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 76498-458
    Pathodorizer (Deodorizer)
    Pathodorizer (Deodorizer)
    1 gallon
    Log in for availability
    Log in for availability
    Each
    Each - $122.39
    $122.39
    Each
       $122.39
    Supplier Number: BE040
  • 76498-460
    Pathodorizer Plus (Deodorizer, cleaner, degreaser)
    Pathodorizer Plus (Deodorizer, cleaner, degreaser)
    1 gallon
    Log in for availability
    Log in for availability
    Each
    Each - $143.13
    $143.13
    Each
       $143.13
    Supplier Number: BE042