Orders LinkContactUsLinkComponent
MicroGenesis Mop
MicroGenesis Mop
Supplier:   Contec
MicroGenesis Mop
Supplier:  Contec
Certificates
MicroGenesis Mop
MicroGenesis Mop
2 Options Available Below
Product Details & Documents
Single-use microfiber mop pads.
  • Synthetic construction is compatible with common disinfectants such as quats, peroxides and bleach
  • New pad for each use simplifies quality control, inventory and space management, all without cross-contamination risk
  • Compatible with most existing hardware
MicroGenesis Mop 100% synthetic, disposable microfiber mop pad effectively removes fine particles and large debris from floors, providing superior removal of visible contaminants. Designed to be used wet or dry for general facilities cleaning in ISO Class 8 environments and above. It is compatible with a wide range of disinfectants, including quats, peroxides and bleach. These synthetic pads are designed to provide uniform surface coverage.
Product Family Options
MicroGenesis Mop Pad
Product Information
  • Size
    Dimensions
    Material
    Availability
    Price
  • 76499-340
    Single-use microfiber pad
    Single-use microfiber pad
    13×130 cm (5×51" )
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $368.08
    $368.08
    New Customer Promotional Price
    Case of 1
       $368.08
    Supplier Number: MGMP0006
Mop Frame
Product Information
  • Size
    Dimensions
    Material
    Availability
    Price
  • 76551-222
    MicroGenesis mop frame, blue
    MicroGenesis mop frame, blue
    10×44 cm (4×17.5")
    PP
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 6
    Case of 6 - $193.50
    $193.50
    New Customer Promotional Price
    Log in for availability
    Each
    Each - $117.23
    $117.23
    New Customer Promotional Price
    Case of 6
       $193.50
    Each
       $117.23
    Supplier Number: MGMP0009
  • 76499-342
    Mop Frame with handle and hook strips
    Mop Frame with handle and hook strips
    107 cm (42") extends to 176.5 cm (69.5")
    Aluminum
    Discontinued
    Supplier Number: -