Orders LinkContactUsLinkComponent
PREempt® Plus Disinfectant Solution, Contec
PREempt® Plus Disinfectant Solution, Contec
Supplier:   Contec
PREempt® Plus Disinfectant Solution, Contec
Supplier:  Contec
Certificates
PREempt® Plus Disinfectant Solution, Contec
PREempt® Plus Disinfectant Solution, Contec
2 Options Available Below
Product Details & Documents
One-step disinfectant cleaner utilizing accelerated Hydrogen Peroxide®.
  • Faster kill times make it easier to maintain compliance and appropriate dwell time
  • Less residue makes it more suitable for critical environments
  • Contains low levels of hydrogen peroxide and nonhazardous, allowing for cost-effective disposal
  • Lowest EPA toxicity rating requires no PPE, contains no VOCs, and is odor-free for better operator acceptability
  • Compatible with a wide range of materials for less wear on surfaces and equipment
PREempt® Plus is a new and improved formulation of PREempt RTU that features faster kill times and lower residue. PREempt Plus is a ready-to-use, one step broad-spectrum cleaner and disinfectant designed for cleaning and disinfecting hard, non-porous surfaces.
This Utilizes Accelerated Hydrogen Peroxide® (AHP®), a patented synergistic blend of commonly used ingredients, that when combined with low levels of hydrogen peroxide dramatically increase its efficacy and cleaning performance.
PREempt Plus kills viruses in 30 seconds, bacteria in 1 minute, fungi in 30 seconds to 1 minute and TB in 1 minute.
This has lower residue than the original PREempt RTU making it more suitable for critical environments. It contains no volatile organic compounds (VOCs) and has a health hazard rating of zero.
Ideal for cleaning and disinfecting environmental surfaces in cleanrooms and laboratory areas including workstations, fume hoods, equipment and other hard non-porous environmental surfaces.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 76499-140
    PREempt Plus disinfectant with accelerated Hydrogen peroxide, capped
    PREempt Plus disinfectant with accelerated Hydrogen peroxide, capped
    32 oz.(0.95 L)
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $120.40
    $120.40
    New Customer Promotional Price
    Case of 12
       $120.40
    Supplier Number: 2B101
  • 76499-142
    PREempt Plus disinfectant with accelerated Hydrogen peroxide, capped
    PREempt Plus disinfectant with accelerated Hydrogen peroxide, capped
    1 gallon (3.8 L)
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $141.04
    $141.04
    New Customer Promotional Price
    Case of 4
       $141.04
    Supplier Number: 2B105
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Trigger Sprayer for 946.3 ml (32 oz.) Bottle
    Trigger Sprayer for 946.3 ml (32 oz.) Bottle
    89066-160
    Log in for availability
    Case of 12 - $63.73