ContactUsLinkComponent
Innovating Science® Determination of Dissolved Carbon Dioxide Concentration
Innovating Science® Determination of Dissolved Carbon Dioxide Concentration
Catalog # 470347-436
Supplier:  Ward's Science
CAS Number:  
undefined
Innovating Science® Determination of Dissolved Carbon Dioxide Concentration
Catalog # 470347-436
Supplier:  Ward's Science
Supplier Number:  IS9757
CAS Number:  
Specifications
  • Chemical method:
    Titration
  • Description:
    Determination of dissolved CO₂ test kit
  • Environmentally Preferable:
  • Parameter:
    Carbon Dioxide
  • Cat. No.:
    470347-436
  • No. of tests:
    40
Specifications
About this item
Easy way to test water for carbon dioxide concentration.
  • 40 water tests for carbon dioxide concentration
The proper amount of dissolved carbon dioxide in water is critical for maintaining a healthy aquatic ecosystem. All aquatic life has an impact on and is in turn impacted by the dissolved carbon dioxide concentration in a system, so tracking the concentration and changes in concentration can help scientists to better understand the health of an aquatic system and the organisms that exist within it.
Delivery: Includes 3 plastic pipettes; 3 reaction vessels; 3 collection bottles; 1 measuring tube; 1 titrator; 15 ml phenolphthalein indicator; 50 ml potassium hydroxide reagent.