Concanavalin A (from Canavalia ensiformis)
Carbohydrate-binding protein that agglutinates various animal cells. Also known to restore the growth pattern of tumor cells to that of normal cells. T-lymphocyte activator.
- CAS No.:11028-71-0
- Source:Canavalia ensiformis
- Form:Lyophilized solid
- Protein/Peptide Name:Concavalin
Frequently Bought Together