VWR® Advanced Binocular Microscope
Catalog # 470331-644
Supplier: VWR International
Specifications
- Magnification:4×, 10×, 40× and 100×
- Description:Binocular microscope, LED, mech stage
- Cat. No.:470331-644
Specifications
About this item
Offers the versatility and precision necessary for research and clinical applications.
- Siedentopf binocular head
- Parfocal and parcentered optics
- Coaxial coarse and fine adjustment controls
- Three-year manufacturer's warranty
Multiple features make this a practical choice for classroom and clinical environments. The combination of Cool-white LED lighting and glass-coated optics provide crisp and clear images allowing students to view more details and minute structures.
Delivery: Microscope includes a manual and a dust cover.