VWR® Standard / VWR® Advanced Standard and Advanced Compound Microscopes
Supplier: VWR International
Certificates
About this item
Quality microscopes at an affordable price.
- Parfocal and parcentered optics
- Separate coarse and fine adjustment controls
- Three-year manufacturer's warranty
These durable microscopes feature all-metal bodies and all-glass-coated optics. They provide crisp, clear images allowing students to view more details and minute structures.
Delivery: All microscopes include a manual and a dust cover.
Frequently Bought Together