Orum International AIRBIO ONE HEPA Filter Kit, Contact Plate, Hardy Diagnostics
76470-200
- Description:Sampler
- Cat. No.:76470-200
- Airflow:100 L/min
- Cartridge type:Contact plate
A benchtop stable viable particle air sampling instrument with an attached HEPA filtration system which filters the exhausted air.
- Features Bluetooth data transfer capability
If viable air monitoring plan involves transferring the air sampler from different ISO classification environments with varied levels of microbial contamination (ISO Class 7 to ISP Class 5, for instance), a HEPA filter equipped TRIO.BAS™ instrument may be used to avoid transfer of particles from the inside of the aspirating chamber.
: One air sampler with power charger, one stainless steel aspirating head with cover, one data transfer cable, a calibration certificate, IQ, OQ, PQ fillable guideline documents, protective carrying case, and one box (10 packs) replacement HEPA filters.