Trace Evidence Envelopes
Trace evidence folds
Catalog # 470334-384
Supplier: FORENSICS SOURCE
Supplier #: 1005341
Jump to:
Product Details & Documents
Perfect for small samples.
- Samples won't escape through corners
- Printed for recording
Collect and transport trace evidence securely with these Trace Evidence Folds. Trace evidence is placed in the center of the fold. Seal using standard evidence tape. There are no corners or creases for minute evidence to escape. To open, simply cut the seal and the Trace Evidence Fold opens into a flat sheet with collected evidence held securely in the center. Each fold is pre-printed with basic chain-of-custody information.
Specifications
- Description:Trace evidence folds