ContactUsLinkComponent
Buccal Swab Collection Kit
Buccal Swab Collection Kit
Catalog # 470334-370
Supplier:  FORENSICS SOURCE
undefined
Buccal Swab Collection Kit
Catalog # 470334-370
Supplier:  FORENSICS SOURCE
Supplier Number:  4-4980
Specifications
  • Description:
    Buccal swab collection kit
  • Cat. No.:
    470334-370
Specifications
About this item
Easy set for field DNA collection.
  • Includes documentation forms and evidence seals
Used to collect Buccal cells for DNA processing. Simply follow the included directions to collect a sample by rubbing the included swabs inside the subject’s mouth. Each kit includes gloves, documentation forms and security labels. Place the swab in the inner envelope, seal it and send to your lab.