ContactUsLinkComponent
Continuous Scale Evidence Tape
Continuous Scale Evidence Tape
Catalog # 470334-320
Supplier:  FORENSICS SOURCE
undefined
Continuous Scale Evidence Tape
Catalog # 470334-320
Supplier:  FORENSICS SOURCE
Supplier Number:  6-3855
Specifications
  • Description:
    Continuous scale evidence tape
  • Environmentally Preferable:
  • Cat. No.:
    470334-320
Specifications
About this item
Photo scale for any surface.
  • Measures millimeters on one edge and inches on the other
  • Apply and remove with no item damage
This adhesive photo evidence scale measures millimeters on one edge and inches on the other. Each 2.5 cm (1") by 9.1 m (30') roll features a repeating pattern every 30.5 cm (12"). The adhesive is non-permanent so that you can apply the scale and remove it without damaging the item. Alternating black bands throughout the scale provide better visualization in distance photography.