Tamper Guard® Evidence Tape
Catalog # 470334-306
Supplier: FORENSICS SOURCE
Specifications
- Description:Evidence tape
- Length:85'
- Environmentally Preferable:
- Cat. No.:470334-306
- Width:1.25"
Specifications
About this item
Makes any container tamperproof.
- Easily secures to containers
- Shows evidence of physical or chemical tampering
- Write-on
Virtually impossible to violate, TAMPER GUARD® Evidence Tape outperforms all evidence tapes and is ideal for anchoring and sealing today’s exotic plastics and other finishes. Each roll is printed in safety orange using a special dye that will bleed and smear if it comes into contact with most solvents, and is printed on a thin acrylic stock that tears easily. A superior adhesive and minute saw-tooth edges along both sides of the tape leave signs of tampering. Easy to write on, this tape is ideal for sealing evidence or for secure storage of nearly any item.