Order Entry
United States
ContactUsLinkComponent
Hook Collar Clamp
Hook Collar Clamp
Catalog # 470325-666
undefined
Hook Collar Clamp
Catalog # 470325-666
Supplier Number:  HKCL01

Specifications

  • Description:
    Hook collar clamp for rods to 13 mm
  • Cat. No.:
    470325-666

Specifications

About this item

Hook Collar clamps are ideal for suspending pulleys, balances, etc. from support rods or cross members.

  • Available for rods up to 0.5" or 0.75"

Features thumbscrews and hooks for suspending pulleys, balances, etc. from support rods or crossbars.