To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Pierce™ 6xHis Protein Tag Stain Reagent Set
- Size:1 l
- Environmentally Preferable:
- Cat. No.:PI24570
- Supplier no.:24570
Specifications
About this item
The Thermo Scientific Pierce 6xHis Protein Tag Stain Reagent Set is a fluorescent stain for preferential detection of histidine-tagged (His-tagged) recombinant fusion proteins in protein polyacrylamide gels.
- Two- to three-times faster than Western blotting—get results sooner and save hours of valuable research time
- Detects directly on the gel—can eliminate the need for a membrane transfer and Western blot step for the detection of at least 0.2 µg of 6xHis-tagged protein
- Ready-to-use, two-reagent formula—no mixing, no diluting and no fuss; guarantees a simple-to-perform, mistake-free detection protocol
- Specific fluorescent detection of 6xHis-tagged proteins—see only what you want to see (CCD camera recommended for low abundance proteins; UV-transilluminator for abundant proteins)
- Compatible with double-staining—follow His-tagged protein staining with Thermo Scientific GelCode Blue Stain Reagent for a total protein profile determination
The unique fluorescent stain specifically detects histidine-tagged fusion proteins that have been electrophoresed in polyacrylamide gels (e.g., SDS-PAGE). With this stain, expression of His-tagged recombinant proteins can be assessed directly in-gel, eliminating membrane transfer and Western blotting steps that are typically used to confirm purity and yield of tagged proteins. Although the lower limit of detection depends on which UV lamp (300nm) and detection equipment (CCD camera best) are used, the stain is convenient for in-process assessment of His-tagged protein production and is compatible with subsequent staining with coomassie dye and other total protein stains.
Includes: Stain and developer solutions (500 mL each)
Requires: UV lamp source (300nm) and CCD camera