Specifications
- Description:Diversey™ Divoflow TC 86 Cleaner
- Size:55 gal.
- Cat. No.:76492-698
Specifications
About this item
Low foam chlorinated detergent for hard water.
- Highly efficient in the removal of organic material including food soil, stains and off-flavors
- Low foaming under conditions of high turbulence
- Good hard water control and free rinsing, leaves no hard water films, minimizes acid cleaning cycles in most clean-in-place (CIP) operations
- Free-rinsing, minimizes water usage
- For use in hard water
Divoflow TC86 cleaner contains a blend of caustic alkali, sodium hypochlorite and organic sequestrants/threshold agents. This combination of ingredients provides excellent soil and stain removal properties for use in hard water conditions. It is recommended as a single-stage cleaner for use in a wide range of CIP and spray washing applications. This product is used in brewing and beverage plants for cleaning fillers, sugar/syrup tanks, open vessels, yeast handling and filtration equipment. It can be used in bars and restaurants for beer line cleaning. It is also used in dairies for cleaning milk reception areas and in food processors for general CIP of equipment and drain cleaning.
This product is low foaming and is suitable for use with a variety of CIP and spray washing equipment.
Use at concentrations between 1.6–32 mL/L (0.2–4 U.S. fl. oz / U.S. gal) depending on the type and degree of soiling.