Order Entry
United States
Orders LinkContactUsLinkComponent
HOBO® Temperature Data Logger

HOBO® Temperature Data Logger

Temperature data logger, –20...70 °C, 0.024 °C at 25 °C, ±0.21 °C from 0...50 °C
Catalog # 470323-520
Supplier #:  UX100-001
HOBO® Temperature Data Logger
Catalog # 470323-520
HOBO® Temperature Data Logger
HOBO® Temperature Data Logger

Temperature data logger, –20...70 °C, 0.024 °C at 25 °C, ±0.21 °C from 0...50 °C

Price
$126.05
Log in for availability
Each
Product Details & Documents

The HOBO® temperature data logger records temperature in indoor environments with its integrated sensor.

  • Easy-to view LCD display
  • Large memory capacity
  • Visual high and low alarm thresholds
  • New Burst and Statistics logging modes
  • Compatible with HOBOware and HOBOware Pro software for logger setup, graphing and analysis

The logger is ideal for monitoring occupant comfort in office buildings, tracking food storage conditions in warehouses, and logging temperature trends in server rooms.

Ordering information: HOBO data loggers require HOBOware software and a USB cable. (Note: A USB cable is included with HOBOware Pro CD).

Specifications
  • Resolution:
    0.024 °C at 25 °C
  • Description:
    Temperature data logger
  • Accuracy:
    ±0.21 °C from 0...50 °C
  • Range:
    –20...70 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.